Anti-CXCR5 Mouse Monoclonal Antibody (Phycoerythrin) [clone: 51505]

Supplier: Novus Biologicals
FAB190P-100 FAB190P-025
103251-788EA 634.86 USD
103251-788 103251-786
Anti-CXCR5 Mouse Monoclonal Antibody (Phycoerythrin) [clone: 51505]
Antibodies
Type: Primary
Antigen: CXCR5
Clonality: Monoclonal
Clone: 51505
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR