Anti-ITGAM Mouse Monoclonal Antibody (Alexa Fluor 647) [clone: 238446]

Supplier: Novus Biologicals
FAB16991R-025 FAB16991R-100
103251-624EA 338.46 USD
103251-624 103251-622
Anti-ITGAM Mouse Monoclonal Antibody (Alexa Fluor 647) [clone: 238446]
Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: 238446
Conjugation: Alexa Fluor 647
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR