Anti-CXCR3 Rat Monoclonal Antibody (Alexa Fluor 700) [clone: 220803]

Supplier: Novus Biologicals
FAB1685N-100 FAB1685N-025
103251-580EA 436.92 USD
103251-580 103251-578
Anti-CXCR3 Rat Monoclonal Antibody (Alexa Fluor 700) [clone: 220803]
Type: Primary
Antigen: CXCR3
Clonality: Monoclonal
Clone: 220803
Conjugation: Alexa Fluor 700
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR