Anti-PODXL Mouse Monoclonal Antibody (Alexa Fluor 700) [clone: 222328]

Supplier: Novus Biologicals
FAB1658N
103251-532EA 466.15 USD
103251-532
Anti-PODXL Mouse Monoclonal Antibody (Alexa Fluor 700) [clone: 222328]
Type: Primary
Antigen: PODXL
Clonality: Monoclonal
Clone: 222328
Conjugation: Alexa Fluor 700
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR