Anti-PODXL Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 222328]

Supplier: Novus Biologicals
FAB1658G
103251-530EA 591.48 USD
103251-530
Anti-PODXL Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 222328]
Antibodies
Type: Primary
Antigen: PODXL
Clonality: Monoclonal
Clone: 222328
Conjugation: Alexa Fluor 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR