Anti-ULBP1 Mouse Monoclonal Antibody (PerCP (Peridinin-chlorophyll Protein Complex)) [clone: 170818]

Supplier: Novus Biologicals
FAB1380C
103251-458EA 663.78 USD
103251-458
Anti-ULBP1 Mouse Monoclonal Antibody (PerCP (Peridinin-chlorophyll Protein Complex)) [clone: 170818]
Antibodies
Type: Primary
Antigen: ULBP1
Clonality: Monoclonal
Clone: 170818
Conjugation: PerCP (Peridinin-chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR