Anti-O4 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: O4]

Supplier: Novus Biologicals
FAB1326G
103251-420EA 663.78 USD
103251-420
Anti-O4 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: O4]
Antibodies
Type: Primary
Antigen: O4
Clonality: Monoclonal
Clone: O4
Conjugation: Alexa Fluor 488
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Chicken, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR