Anti-MICA Mouse Monoclonal Antibody (Alexa Fluor 700) [clone: 159207]

Supplier: Novus Biologicals
FAB13001N-025 FAB13001N-100
103251-412EA 591.48 USD
103251-412 103251-410
Anti-MICA Mouse Monoclonal Antibody (Alexa Fluor 700) [clone: 159207]
Antibodies
Type: Primary
Antigen: MICA
Clonality: Monoclonal
Clone: 159207
Conjugation: Alexa Fluor 700
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR