Anti-CD40LG Rat Monoclonal Antibody (Fluorescein) [clone: 208109]

Supplier: Novus Biologicals
FAB1163F
103251-324EA 295.38 USD
103251-324
Anti-CD40LG Rat Monoclonal Antibody (Fluorescein) [clone: 208109]
Type: Primary
Antigen: CD40LG
Clonality: Monoclonal
Clone: 208109
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR