Anti-CXCR3 Mouse Monoclonal Antibody (Phycoerythrin) [clone: 49801]

Supplier: Novus Biologicals
FAB160P-025 FAB160P-100
103251-286EA 591.48 USD
103251-286 103251-284
Anti-CXCR3 Mouse Monoclonal Antibody (Phycoerythrin) [clone: 49801]
Antibodies
Type: Primary
Antigen: CXCR3
Clonality: Monoclonal
Clone: 49801
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR