Anti-CD163 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 215927]

Supplier: Novus Biologicals
FAB1607A-100 FAB1607A-025
103251-236EA 533.63 USD
103251-236 103251-234
Anti-CD163 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 215927]
Antibodies
Type: Primary
Antigen: CD163
Clonality: Monoclonal
Clone: 215927
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR