Anti-CCR4 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 205410]

Supplier: Novus Biologicals
FAB1567G-025
103251-170EA 258.86 USD
103251-170
Anti-CCR4 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 205410]
Antibodies
Type: Primary
Antigen: CCR4
Clonality: Monoclonal
Clone: 205410
Conjugation: Alexa Fluor 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR