Anti-CD274 Mouse Monoclonal Antibody (PerCP (Peridinin-chlorophyll Protein Complex)) [clone: 130021]

Supplier: Novus Biologicals
FAB1561C-025 FAB1561C-100
103251-126EA 605.94 USD
103251-126 103251-124
Anti-CD274 Mouse Monoclonal Antibody (PerCP (Peridinin-chlorophyll Protein Complex)) [clone: 130021]
Antibodies
Type: Primary
Antigen: CD274
Clonality: Monoclonal
Clone: 130021
Conjugation: PerCP (Peridinin-chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR