Anti-CCR3 Rat Monoclonal Antibody (Fluorescein) [clone: 61828]

Supplier: Novus Biologicals
FAB155F-100 FAB155F-025
103251-098EA 461.32 USD
103251-098 103251-096
Anti-CCR3 Rat Monoclonal Antibody (Fluorescein) [clone: 61828]
Antibodies
Type: Primary
Antigen: CCR3
Clonality: Monoclonal
Clone: 61828
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR