Anti-KLRK1 Rat Monoclonal Antibody (Fluorescein) [clone: 191004]

Supplier: Novus Biologicals
FAB1547F-100 FAB1547F-025
103251-018EA 309.23 USD
103251-018 103251-016
Anti-KLRK1 Rat Monoclonal Antibody (Fluorescein) [clone: 191004]
Type: Primary
Antigen: KLRK1
Clonality: Monoclonal
Clone: 191004
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR