Anti-Tdgf1 Rat Monoclonal Antibody (Allophycocyanin) [clone: 237603]

Supplier: Novus Biologicals
FAB1538A
103250-974EA 605.94 USD
103250-974
Anti-Tdgf1 Rat Monoclonal Antibody (Allophycocyanin) [clone: 237603]
Antibodies
Type: Primary
Antigen: Tdgf1
Clonality: Monoclonal
Clone: 237603
Conjugation: Allophycocyanin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR