Anti-CD8A Mouse Monoclonal Antibody (Fluorescein) [clone: 37006]

Supplier: Novus Biologicals
FAB1509F-100 FAB1509F-025
103250-914EA 172.09 USD
103250-914 103250-916
Anti-CD8A Mouse Monoclonal Antibody (Fluorescein) [clone: 37006]
Antibodies
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 37006
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR