Anti-DLL4 Rat Monoclonal Antibody (Alexa Fluor 647) [clone: 447506]

Supplier: Novus Biologicals
FAB1506R-025
103250-896EA 224.62 USD
103250-896
Anti-DLL4 Rat Monoclonal Antibody (Alexa Fluor 647) [clone: 447506]
Type: Primary
Antigen: DLL4
Clonality: Monoclonal
Clone: 447506
Conjugation: Alexa Fluor 647
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR