Anti-SIGLEC5 Mouse Monoclonal Antibody (Fluorescein) [clone: 194128]

Supplier: Novus Biologicals
FAB10721F
103250-674EA 352.31 USD
103250-674
Anti-SIGLEC5 Mouse Monoclonal Antibody (Fluorescein) [clone: 194128]
Type: Primary
Antigen: SIGLEC5
Clonality: Monoclonal
Clone: 194128
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR