Anti-PDGFRA Goat Polyclonal Antibody (Allophycocyanin)

Supplier: Novus Biologicals
FAB1062A
103250-666EA 663.78 USD
103250-666
Anti-PDGFRA Goat Polyclonal Antibody (Allophycocyanin)
Antibodies
Type: Primary
Antigen: PDGFRA
Clonality: Polyclonal
Clone:
Conjugation: Allophycocyanin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR