Anti-TREM1 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 193015]

Supplier: Novus Biologicals
FAB1278A
103250-546EA 605.94 USD
103250-546
Anti-TREM1 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 193015]
Antibodies
Type: Primary
Antigen: TREM1
Clonality: Monoclonal
Clone: 193015
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR