Anti-FCGR1A Mouse Monoclonal Antibody (Fluorescein) [clone: 276426]

Supplier: Novus Biologicals
FAB12571F
103250-484EA 267.69 USD
103250-484
Anti-FCGR1A Mouse Monoclonal Antibody (Fluorescein) [clone: 276426]
Type: Primary
Antigen: FCGR1A
Clonality: Monoclonal
Clone: 276426
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR