Anti-TNFSF4 Goat Polyclonal Antibody (Phycoerythrin)

Supplier: Novus Biologicals
FAB1236P
103250-466EA 504.71 USD
103250-466
Anti-TNFSF4 Goat Polyclonal Antibody (Phycoerythrin)
Antibodies
Type: Primary
Antigen: TNFSF4
Clonality: Polyclonal
Clone:
Conjugation: Phycoerythrin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR