Anti-LGALS3 Rat Monoclonal Antibody (Alexa Fluor 488) [clone: 202213]

Supplier: Novus Biologicals
FAB1197G
103250-382EA 436.92 USD
103250-382
Anti-LGALS3 Rat Monoclonal Antibody (Alexa Fluor 488) [clone: 202213]
Type: Primary
Antigen: LGALS3
Clonality: Monoclonal
Clone: 202213
Conjugation: Alexa Fluor 488
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR