Anti-CD300A Rat Monoclonal Antibody (Phycoerythrin) [clone: 172224]

Supplier: Novus Biologicals
FAB1186P
103250-324EA 475.78 USD
103250-324
Anti-CD300A Rat Monoclonal Antibody (Phycoerythrin) [clone: 172224]
Antibodies
Type: Primary
Antigen: CD300A
Clonality: Monoclonal
Clone: 172224
Conjugation: Phycoerythrin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR