Anti-ALCAM Goat Polyclonal Antibody (Phycoerythrin)

Supplier: Novus Biologicals
FAB1172P
103250-310EA 533.63 USD
103250-310
Anti-ALCAM Goat Polyclonal Antibody (Phycoerythrin)
Antibodies
Type: Primary
Antigen: ALCAM
Clonality: Polyclonal
Clone:
Conjugation: Phycoerythrin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR