Anti-SIGLEC7 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 194211]

Supplier: Novus Biologicals
FAB11381A-100 FAB11381A-025
103250-190EA 634.86 USD
103250-190 103250-188
Anti-SIGLEC7 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 194211]
Antibodies
Type: Primary
Antigen: SIGLEC7
Clonality: Monoclonal
Clone: 194211
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR