Anti-EGFR Rat Monoclonal Antibody (Phycoerythrin) [clone: 423103]

Supplier: Novus Biologicals
FAB10951P
103250-166EA 548.09 USD
103250-166
Anti-EGFR Rat Monoclonal Antibody (Phycoerythrin) [clone: 423103]
Antibodies
Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 423103
Conjugation: Phycoerythrin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR