Anti-KLRC1 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 131411]

Supplier: Novus Biologicals
FAB1059A-025 FAB1059A-100
103250-132EA 258.86 USD
103250-132 103250-134
Anti-KLRC1 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 131411]
Antibodies
Type: Primary
Antigen: KLRC1
Clonality: Monoclonal
Clone: 131411
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR