Anti-NCR3 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 210845]

Supplier: Novus Biologicals
FAB1849G-025 FAB1849G-100
103246-360EA 562.55 USD
103246-360 103246-358
Anti-NCR3 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 210845]
Antibodies
Type: Primary
Antigen: NCR3
Clonality: Monoclonal
Clone: 210845
Conjugation: Alexa Fluor 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR