Anti-CCR5 Mouse Monoclonal Antibody (Fluorescein) [clone: 45531]

Supplier: Novus Biologicals
FAB182F-100 FAB182F-025
103246-252EA 186.55 USD
103246-252 103246-254
Anti-CCR5 Mouse Monoclonal Antibody (Fluorescein) [clone: 45531]
Antibodies
Type: Primary
Antigen: CCR5
Clonality: Monoclonal
Clone: 45531
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR