Anti-CCR9 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 112509]

Supplier: Novus Biologicals
FAB179A-100 FAB179A-025
103246-196EA 229.94 USD
103246-196 103246-198
Anti-CCR9 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 112509]
Antibodies
Type: Primary
Antigen: CCR9
Clonality: Monoclonal
Clone: 112509
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR