Anti-CD83 Mouse Monoclonal Antibody (Fluorescein) [clone: HB15e]

Supplier: Novus Biologicals
FAB1774F
103245-950EA 475.78 USD
103245-950
Anti-CD83 Mouse Monoclonal Antibody (Fluorescein) [clone: HB15e]
Antibodies
Type: Primary
Antigen: CD83
Clonality: Monoclonal
Clone: HB15e
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR