Anti-Raet1a Rat Monoclonal Antibody (Alexa Fluor 488) [clone: 199215]

Supplier: Novus Biologicals
FAB1758G
103245-934EA 436.92 USD
103245-934
Anti-Raet1a Rat Monoclonal Antibody (Alexa Fluor 488) [clone: 199215]
Type: Primary
Antigen: Raet1a
Clonality: Monoclonal
Clone: 199215
Conjugation: Alexa Fluor 488
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR