Anti-CLEC7A Rat Monoclonal Antibody (Alexa Fluor 700) [clone: 218820]

Supplier: Novus Biologicals
FAB17561N
103245-914EA 533.63 USD
103245-914
Anti-CLEC7A Rat Monoclonal Antibody (Alexa Fluor 700) [clone: 218820]
Antibodies
Type: Primary
Antigen: CLEC7A
Clonality: Monoclonal
Clone: 218820
Conjugation: Alexa Fluor 700
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR