Anti-CXCR4 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 44717]

Supplier: Novus Biologicals
FAB173A-100 FAB173A-025
103245-752EA 605.94 USD
103245-752 103245-750
Anti-CXCR4 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 44717]
Antibodies
Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 44717
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR