Anti-CXCR4 Mouse Monoclonal Antibody (Alexa Fluor 700) [clone: 12G5]

Supplier: Novus Biologicals
FAB170N-100 FAB170N-025
103245-702EA 118 USD
103245-702 103245-704
Anti-CXCR4 Mouse Monoclonal Antibody (Alexa Fluor 700) [clone: 12G5]
Antibodies
Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 12G5
Conjugation: Alexa Fluor 700
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR