Anti-CRTAM Mouse Monoclonal Antibody (Allophycocyanin) [clone: 210213]

Supplier: Novus Biologicals
FAB16951A
103245-634EA 620.4 USD
103245-634
Anti-CRTAM Mouse Monoclonal Antibody (Allophycocyanin) [clone: 210213]
Antibodies
Type: Primary
Antigen: CRTAM
Clonality: Monoclonal
Clone: 210213
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR