Anti-CD209 Mouse Monoclonal Antibody (Fluorescein) [clone: 120612]

Supplier: Novus Biologicals
FAB1621F
103245-538EA 338.46 USD
103245-538
Anti-CD209 Mouse Monoclonal Antibody (Fluorescein) [clone: 120612]
Type: Primary
Antigen: CD209
Clonality: Monoclonal
Clone: 120612
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR