Anti-CCR2 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 48607]

Supplier: Novus Biologicals
FAB151G-100 FAB151G-025
103245-500EA 634.86 USD
103245-500 103245-498
Anti-CCR2 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 48607]
Antibodies
Type: Primary
Antigen: CCR2
Clonality: Monoclonal
Clone: 48607
Conjugation: Alexa Fluor 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR