Anti-ULBP3 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 166510]

Supplier: Novus Biologicals
FAB1517A
103245-470EA 663.78 USD
103245-470
Anti-ULBP3 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 166510]
Antibodies
Type: Primary
Antigen: ULBP3
Clonality: Monoclonal
Clone: 166510
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR