Anti-TLR1 Goat Polyclonal Antibody (Allophycocyanin)

Supplier: Novus Biologicals
FAB1484A
103245-436EA 605.94 USD
103245-436
Anti-TLR1 Goat Polyclonal Antibody (Allophycocyanin)
Antibodies
Type: Primary
Antigen: TLR1
Clonality: Polyclonal
Clone:
Conjugation: Allophycocyanin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR