Anti-IL27RA Mouse Monoclonal Antibody (Phycoerythrin) [clone: 191106]

Supplier: Novus Biologicals
FAB14791P-100 FAB14791P-025
103245-416EA 591.48 USD
103245-416 103245-414
Anti-IL27RA Mouse Monoclonal Antibody (Phycoerythrin) [clone: 191106]
Antibodies
Type: Primary
Antigen: IL27RA
Clonality: Monoclonal
Clone: 191106
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR