Anti-CD86 Mouse Monoclonal Antibody (Fluorescein) [clone: 37301]

Supplier: Novus Biologicals
FAB141F-025 FAB141F-100
103245-368EA 244.4 USD
103245-368 103245-370
Anti-CD86 Mouse Monoclonal Antibody (Fluorescein) [clone: 37301]
Antibodies
Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: 37301
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR