Anti-CD86 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 37301]

Supplier: Novus Biologicals
FAB141A-100 FAB141A-025
103245-362EA 504.71 USD
103245-362 103245-360
Anti-CD86 Mouse Monoclonal Antibody (Allophycocyanin) [clone: 37301]
Antibodies
Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: 37301
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR