Anti-SLC2A1 Mouse Monoclonal Antibody (Alexa Fluor (R) 488) [clone: 202915]

Supplier: Novus Biologicals
FAB1418G
103245-344EA 466.15 USD
103245-344
Anti-SLC2A1 Mouse Monoclonal Antibody (Alexa Fluor (R) 488) [clone: 202915]
Type: Primary
Antigen: SLC2A1
Clonality: Monoclonal
Clone: 202915
Conjugation: Alexa Fluor (R) 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR