Anti-KLRC2 Mouse Monoclonal Antibody (Alexa Fluor 647) [clone: 134591]

Supplier: Novus Biologicals
FAB138R-025
103245-296EA 258.86 USD
103245-296
Anti-KLRC2 Mouse Monoclonal Antibody (Alexa Fluor 647) [clone: 134591]
Antibodies
Type: Primary
Antigen: KLRC2
Clonality: Monoclonal
Clone: 134591
Conjugation: Alexa Fluor 647
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR