Anti-ITGA4 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 7.2R]

Supplier: Novus Biologicals
FAB1354G
103245-196EA 432.4 USD
103245-196
Anti-ITGA4 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 7.2R]
Antibodies
Type: Primary
Antigen: ITGA4
Clonality: Monoclonal
Clone: 7.2R
Conjugation: Alexa Fluor 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR