Anti-ULBP-2/5/6 Mouse Monoclonal Antibody (Phycoerythrin) [clone: 165903]

Supplier: Novus Biologicals
FAB1298P
103245-146EA 663.78 USD
103245-146
Anti-ULBP-2/5/6 Mouse Monoclonal Antibody (Phycoerythrin) [clone: 165903]
Antibodies
Type: Primary
Antigen: ULBP-2/5/6
Clonality: Monoclonal
Clone: 165903
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR