Anti-GYPA Mouse Monoclonal Antibody (Phycoerythrin) [clone: R10]

Supplier: Novus Biologicals
FAB12281P
103245-062EA 533.63 USD
103245-062
Anti-GYPA Mouse Monoclonal Antibody (Phycoerythrin) [clone: R10]
Antibodies
Type: Primary
Antigen: GYPA
Clonality: Monoclonal
Clone: R10
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR